Assay Details
| Gene Symbol: CYLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100435.2 | XP_014955921.1 | 3 - 4 | 1014 | 71 | |
| XM_015100436.2 | XP_014955922.1 | 3 - 4 | 1037 | 71 | ||
| XM_015100438.2 | XP_014955924.1 | 2 - 3 | 354 | 71 | ||
| XM_027977829.1 | XP_027833630.1 | 2 - 3 | 3159 | 71 | ||
| XM_027977831.1 | XP_027833632.1 | 3 - 4 | 1161 | 71 | ||
More Information
Set Membership: |
|

CYLD
Genomic Map