Assay Details
| Gene Symbol: ABHD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018359.3 | XP_004018408.1 | 4 - 5 | 532 | 56 | |
| XM_012099816.3 | XP_011955206.1 | 4 - 5 | 488 | 56 | ||
| XM_012099819.3 | XP_011955209.1 | 3 - 4 | 428 | 56 | ||
| XM_012099820.3 | XP_011955210.1 | 3 - 4 | 457 | 56 | ||
| XM_027958245.1 | XP_027814046.1 | 4 - 5 | 876 | 56 | ||
More Information
Set Membership: |
|

ABHD6
Genomic Map