Assay Details
| Gene Symbol: CIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114765.1 | NP_001108237.1 | 4 - 5 | 387 | 72 | |
| XM_012098269.3 | XP_011953659.1 | 4 - 5 | 520 | 72 | ||
| GenBank mRNA | EU430744.1 | - | 4 - 5 | 387 | 72 | |
More Information
Set Membership: |
|

CIB1
Genomic Map