Assay Details
| Gene Symbol: SUPT7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005776.4 | XP_004005825.1 | 2 - 3 | 865 | 65 | |
| XM_012166878.3 | XP_012022268.1 | 3 - 4 | 943 | 65 | ||
More Information
Set Membership: |
|

SUPT7L
Genomic Map