Assay Details
| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958487.1 | XP_027814288.1 | 2 - 3 | 1355 | 69 | |
| XM_027958488.1 | XP_027814289.1 | 3 - 4 | 1177 | 69 | ||
| XM_027958489.1 | XP_027814290.1 | 2 - 3 | 1352 | 69 | ||
More Information
Set Membership: |
|

MAD2L1BP
Genomic Map