Assay Details
| Gene Symbol: TM2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970712.1 | XP_027826513.1 | 4 - 5 | 451 | 68 | |
| XM_027970713.1 | XP_027826514.1 | 4 - 5 | 451 | 68 | ||
| XM_027970715.1 | XP_027826516.1 | 4 - 5 | 451 | 68 | ||
More Information
Set Membership: |
|

TM2D1
Genomic Map