Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012149891.3 | XP_012005281.2 | 7 - 8 | 1358 | 81 | |
| XM_012149905.3 | XP_012005295.2 | 6 - 7 | 1588 | 81 | ||
| XM_027965380.1 | XP_027821181.1 | 7 - 8 | 1436 | 81 | ||
| GenBank mRNA | AF499612.1 | - | 2 - 3 | 244 | 81 | |
More Information
Set Membership: |
|

HCRTR1
Genomic Map