Assay Details
| Gene Symbol: SRI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163603.1 | NP_001157075.1 | 4 - 5 | 271 | 62 | |
| GenBank mRNA | FJ937948.1 | - | 4 - 5 | 271 | 62 | |
More Information
Set Membership: |
|

SRI
Genomic Map