Assay Details
| Gene Symbol: SPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287469.1 | NP_001274398.1 | 3 - 4 | 184 | 64 | |
| XM_027965740.1 | XP_027821541.1 | 2 - 3 | 409 | 64 | ||
| GenBank mRNA | KF408269.1 | - | 3 - 4 | 184 | 64 | |
More Information
Set Membership: |
|

SPX
Genomic Map