Assay Details
| Gene Symbol: IMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964659.1 | XP_027820460.1 | 5 - 6 | 528 | 56 | |
| XM_027964660.1 | XP_027820461.1 | 5 - 6 | 1100 | 56 | ||
More Information
Set Membership: |
|

IMP4
Genomic Map