Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103192.2 | XP_014958678.2 | 2 - 3 | 383 | 78 | |
| XM_027959755.1 | XP_027815556.1 | 2 - 3 | 373 | 78 | ||
More Information
Set Membership: |
|

GAL3ST3
Genomic Map