Assay Details
| Gene Symbol: MMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961497.1 | XP_027817298.1 | 3 - 4 | 335 | 64 | |
| XM_027961498.1 | XP_027817299.1 | 3 - 4 | 326 | 64 | ||
| XM_027961499.1 | XP_027817300.1 | 3 - 4 | 326 | 64 | ||
More Information
Set Membership: |
|

MMD2
Genomic Map