Assay Details
| Gene Symbol: LOC101102529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961780.1 | XP_027817581.1 | 20 - 21 | 2096 | 101 | |
| XM_027961781.1 | XP_027817582.1 | 20 - 21 | 2096 | 101 | ||
| XM_027961782.1 | XP_027817583.1 | 20 - 21 | 2096 | 101 | ||
| XM_027961783.1 | XP_027817584.1 | 14 - 15 | 1533 | 101 | ||
More Information
Set Membership: |
|

LOC101102529
Genomic Map