Assay Details
| Gene Symbol: SPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012788.4 | XP_004012837.1 | 5 - 6 | 645 | 64 | |
| XM_027974688.1 | XP_027830489.1 | 4 - 5 | 671 | 64 | ||
More Information
Set Membership: |
|

SPOP
Genomic Map