Assay Details
| Gene Symbol: LOC101108428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018389.4 | XP_004018438.1 | 2 - 3 | 281 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101108428| Gene Symbol: LOC101108428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018389.4 | XP_004018438.1 | 2 - 3 | 281 | 80 | |