Assay Details
| Gene Symbol: PIP4K2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006533.4 | XP_004006582.2 | 5 - 6 | 717 | 71 | |
| XM_012174662.3 | XP_012030052.2 | 5 - 6 | 716 | 71 | ||
| XM_012174663.3 | XP_012030053.2 | 5 - 6 | 717 | 71 | ||
More Information
Set Membership: |
|

PIP4K2C
Genomic Map