Assay Details
| Gene Symbol: CRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975500.1 | XP_027831301.1 | 12 - 13 | 1802 | 68 | |
| XM_027975502.1 | XP_027831303.1 | 12 - 13 | 1787 | 68 | ||
| XM_027975506.1 | XP_027831307.1 | 11 - 12 | 1733 | 68 | ||
| XM_027975510.1 | XP_027831311.1 | 11 - 12 | 1718 | 68 | ||
| XM_027975514.1 | XP_027831315.1 | 12 - 13 | 1691 | 68 | ||
More Information
Set Membership: |
|

CRTC2
Genomic Map