Assay Details
| Gene Symbol: LOC101117622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102048.2 | XP_014957534.2 | 14 - 15 | 3002 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117622| Gene Symbol: LOC101117622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102048.2 | XP_014957534.2 | 14 - 15 | 3002 | 60 | |
Set Membership: |
|