Assay Details
| Gene Symbol: LOC101113487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007023.3 | XP_004007072.1 | 10 - 11 | 1127 | 72 | |
| XM_012175713.2 | XP_012031103.1 | 10 - 11 | 1124 | 72 | ||
| XM_012175715.3 | XP_012031105.1 | 10 - 11 | 1120 | 72 | ||
More Information
Set Membership: |
|

LOC101113487
Genomic Map