Assay Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099038.3 | XP_011954428.2 | 1 - 2 | 4911 | 53 | |
| XM_012099040.3 | XP_011954430.2 | 3 - 4 | 440 | 53 | ||
| XM_012099041.3 | XP_011954431.2 | 4 - 5 | 497 | 53 | ||
| XM_015102051.2 | XP_014957537.2 | 2 - 3 | 3068 | 53 | ||
| XM_027957398.1 | XP_027813199.1 | 2 - 3 | 4792 | 53 | ||
More Information
Set Membership: |
|

SLC25A29
Genomic Map