Assay Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961368.1 | XP_027817169.1 | 1 - 2 | 2524 | 55 | |
| XM_027961369.1 | XP_027817170.1 | 3 - 4 | 383 | 55 | ||
| XM_027961370.1 | XP_027817171.1 | 2 - 3 | 2346 | 55 | ||
More Information
Set Membership: |
|

MCRIP2
Genomic Map