Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974413.1 | XP_027830214.1 | 9 - 10 | 1379 | 58 | |
| XM_027974414.1 | XP_027830215.1 | 8 - 9 | 1266 | 58 | ||
More Information
Set Membership: |
|

GFAP
Genomic Map