Assay Details
| Gene Symbol: RBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960154.1 | XP_027815955.1 | 4 - 5 | 625 | 63 | |
| GenBank mRNA | L11346.1 | - | 3 - 4 | 312 | 63 | |
More Information
Set Membership: |
|

RBP4
Genomic Map