Assay Details
| Gene Symbol: LOC101117350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976415.1 | XP_027832216.1 | 3 - 4 | 378 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117350| Gene Symbol: LOC101117350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976415.1 | XP_027832216.1 | 3 - 4 | 378 | 89 | |