Assay Details
| Gene Symbol: LOC101120030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971210.1 | XP_027827011.1 | 19 - 20 | 2086 | 99 | |
| XM_027971216.1 | XP_027827017.1 | 18 - 19 | 1807 | 99 | ||
More Information
Set Membership: |
|

LOC101120030
Genomic Map