Assay Details
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974091.1 | XP_027829892.1 | 1 - 2 | 123 | 57 | |
XM_027974092.1 | XP_027829893.1 | 1 - 2 | 126 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
