Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176869.3 | XP_012032259.2 | 14 - 15 | 1897 | 57 | |
| XM_012176870.3 | XP_012032260.2 | 14 - 15 | 1897 | 57 | ||
More Information
Set Membership: |
|

MINDY4
Genomic Map