Assay Details
Gene Symbol: DAZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964130.1 | XP_027819931.1 | 5 - 6 | 778 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
