Assay Details
| Gene Symbol: RHOBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105116.3 | XP_011960506.1 | 3 - 4 | 309 | 60 | |
| XM_027962357.1 | XP_027818158.1 | 3 - 4 | 474 | 60 | ||
| XM_027962358.1 | XP_027818159.1 | 2 - 3 | 423 | 60 | ||
| XM_027962359.1 | XP_027818160.1 | 3 - 4 | 474 | 60 | ||
More Information
Set Membership: |
|

RHOBTB1
Genomic Map