Assay Details
| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018939.4 | XP_004018988.2 | 3 - 4 | 624 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAPCD1| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018939.4 | XP_004018988.2 | 3 - 4 | 624 | 79 | |