Assay Details
| Gene Symbol: EXTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964605.1 | XP_027820406.1 | 4 - 5 | 2962 | 61 | |
| XM_027964606.1 | XP_027820407.1 | 6 - 7 | 3163 | 61 | ||
| XM_027964607.1 | XP_027820408.1 | 5 - 6 | 3058 | 61 | ||
| XM_027964608.1 | XP_027820409.1 | 4 - 5 | 2901 | 61 | ||
More Information
Set Membership: |
|

EXTL3
Genomic Map