Assay Details
| Gene Symbol: HNRNPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005133.4 | XP_004005182.2 | 6 - 7 | 785 | 86 | |
| XM_027965539.1 | XP_027821340.1 | 6 - 7 | 781 | 86 | ||
| XM_027965540.1 | XP_027821341.1 | 6 - 7 | 777 | 86 | ||
| XM_027965541.1 | XP_027821342.1 | 6 - 7 | 781 | 86 | ||
| XM_027965542.1 | XP_027821343.1 | 5 - 6 | 637 | 86 | ||
| XM_027965543.1 | XP_027821344.1 | 5 - 6 | 631 | 86 | ||
More Information
Set Membership: |
|

HNRNPR
Genomic Map