Assay Details
| Gene Symbol: CCNI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971121.1 | XP_027826922.1 | 6 - 7 | 1293 | 71 | |
| XM_027971122.1 | XP_027826923.1 | 5 - 6 | 1136 | 71 | ||
More Information
Set Membership: |
|

CCNI
Genomic Map