Assay Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010231.4 | XP_004010280.2 | 4 - 5 | 1297 | 77 | |
| XM_012180735.3 | XP_012036125.2 | 4 - 5 | 1300 | 77 | ||
More Information
Set Membership: |
|

PARP16
Genomic Map