Assay Details
| Gene Symbol: EEFSEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957859.1 | XP_027813660.1 | 4 - 5 | 788 | 67 | |
| XM_027957860.1 | XP_027813661.1 | 4 - 5 | 548 | 67 | ||
| XM_027957861.1 | XP_027813662.1 | 4 - 5 | 550 | 67 | ||
More Information
Set Membership: |
|

EEFSEC
Genomic Map