Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020899.3 | XP_004020948.1 | 3 - 4 | 342 | 69 | |
| XM_012104385.2 | XP_011959775.1 | 3 - 4 | 321 | 69 | ||
| XM_012104387.2 | XP_011959777.1 | 3 - 4 | 343 | 69 | ||
| XM_027961843.1 | XP_027817644.1 | 3 - 4 | 344 | 69 | ||
More Information
Set Membership: |
|

CD2BP2
Genomic Map