Assay Details
| Gene Symbol: GEM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973223.1 | XP_027829024.1 | 3 - 4 | 1512 | 63 | |
| XM_027973224.1 | XP_027829025.1 | 3 - 4 | 484 | 63 | ||
More Information
Set Membership: |
|

GEM
Genomic Map