Assay Details
Gene Symbol: LOC101113696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977945.1 | XP_027833746.1 | 21 - 22 | 3421 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
