Assay Details
| Gene Symbol: STX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961311.1 | XP_027817112.1 | 7 - 8 | 623 | 60 | |
| XM_027961312.1 | XP_027817113.1 | 7 - 8 | 586 | 60 | ||
More Information
Set Membership: |
|

STX1A
Genomic Map