Assay Details
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163605.1 | NP_001157077.1 | 3 - 4 | 364 | 77 | |
XM_015092739.2 | XP_014948225.1 | 4 - 5 | 900 | 77 | ||
XM_027963869.1 | XP_027819670.1 | 4 - 5 | 984 | 77 | ||
GenBank mRNA | FJ937959.1 | - | 3 - 4 | 364 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
