Assay Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974501.1 | XP_027830302.1 | 3 - 4 | 925 | 56 | |
| XM_027974502.1 | XP_027830303.1 | 4 - 5 | 992 | 56 | ||
More Information
Set Membership: |
|

HSD17B1
Genomic Map