Assay Details
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257093.1 | NP_001244022.1 | 3 - 4 | 500 | 60 | |
| XM_012096672.3 | XP_011952062.2 | 3 - 4 | 914 | 60 | ||
| XM_012096675.3 | XP_011952065.2 | 3 - 4 | 918 | 60 | ||
| GenBank mRNA | HM245791.1 | - | 1 - 2 | 56 | 60 | |
| JQ348894.1 | - | 3 - 4 | 500 | 60 | ||
| KF833357.1 | - | 3 - 4 | 599 | 60 | ||
More Information
Set Membership: |
|

FST
Genomic Map