Assay Details
| Gene Symbol: C3H22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006984.4 | XP_004007033.2 | 5 - 6 | 655 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3H22orf23| Gene Symbol: C3H22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006984.4 | XP_004007033.2 | 5 - 6 | 655 | 81 | |