Assay Details
| Gene Symbol: LOC101104372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021933.4 | XP_004021982.2 | 7 - 8 | 907 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104372| Gene Symbol: LOC101104372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021933.4 | XP_004021982.2 | 7 - 8 | 907 | 63 | |
Set Membership: |
|