Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010441.4 | XP_004010490.1 | 2 - 3 | 191 | 63 | |
| GenBank mRNA | EU366491.1 | - | 2 - 3 | 103 | 63 | |
More Information
Set Membership: |
|

SRP14
Genomic Map