Assay Details
| Gene Symbol: PYROXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006797.4 | XP_004006846.1 | 10 - 11 | 1204 | 68 | |
| XM_012175221.3 | XP_012030611.1 | 10 - 11 | 1365 | 68 | ||
| XM_012175222.3 | XP_012030612.1 | 11 - 12 | 1473 | 68 | ||
| XM_027967826.1 | XP_027823627.1 | 6 - 7 | 1715 | 68 | ||
More Information
Set Membership: |
|

PYROXD1
Genomic Map