Assay Details
| Gene Symbol: KIF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189031.3 | XP_012044421.1 | 8 - 9 | 2221 | 55 | |
| XM_012189033.3 | XP_012044423.1 | 7 - 8 | 2091 | 55 | ||
| XM_027977064.1 | XP_027832865.1 | 7 - 8 | 2080 | 55 | ||
More Information
Set Membership: |
|

KIF3B
Genomic Map