Assay Details
| Gene Symbol: BTF3L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004001972.3 | XP_004002021.1 | 4 - 5 | 492 | 97 | |
| XM_004001973.4 | XP_004002022.1 | 4 - 5 | 434 | 97 | ||
| XM_012142126.2 | XP_011997516.1 | 5 - 6 | 474 | 97 | ||
More Information
Set Membership: |
|

BTF3L4
Genomic Map