Assay Details
| Gene Symbol: RCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959703.1 | XP_027815504.1 | 5 - 6 | 637 | 66 | |
| XM_027959704.1 | XP_027815505.1 | 5 - 6 | 637 | 66 | ||
| XM_027959705.1 | XP_027815506.1 | 5 - 6 | 641 | 66 | ||
More Information
Set Membership: |
|

RCE1
Genomic Map