Assay Details
| Gene Symbol: LOC101120386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004709.4 | XP_004004758.1 | 1 - 2 | 421 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120386| Gene Symbol: LOC101120386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004709.4 | XP_004004758.1 | 1 - 2 | 421 | 73 | |
Set Membership: |
|